ContactUsLinkComponent
Griffin Beakers, Polypropylene, Chemglass
Catalogue # 89028-358
Fournisseur:  Chemglass
Fournisseur #: CG-1108-L-02
Small Business Enterprise
undefined
Griffin Beakers, Polypropylene, Chemglass
Catalogue # 89028-358
Fournisseur:  Chemglass
Supplier Number:  CG-1108-L-02
Fournisseur #: CG-1108-L-02
Small Business Enterprise
S'identifier pour la disponibilité
Select an Option
10 L (338.14 oz.)
Select an Option
PP Griffin Beaker, Graduated
Select an Option
89028-358
Select an Option
CG-1108-L-02
À propos de cet article
Large scale graduated beaker with flared top and a pouring spout
Autoclavable.
Frequently Bought Together