Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Volume:10 µl
- Environmentally Preferable:
- Cat. no.:10055-204
- Needle Length:57 mm (21/4")
- Gauge:26s
- Supplier No.:365D0291
Spécifications
À propos de cet article
Achieve accurate, precise, and reproducible injections easily using Thermo Scientific™ GC Syringes for Thermo Scientific™ TriPlus™ RSH Autosampler
The robust, field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes.