Lens Cleaning Towelette, MCR Safety
80081-972
- Description:Lens Cleaning Tissues, 100 per Box
- Cat. no.:80081-972
- Supplier No.:LCT
Anti-fog, anti-static lens cleaning towelettes.
- For plastic, PC and glass lenses
- Contain no silicone
From cases, cords, and everything in between we have your covered for all your eyewear accessories needs. Be accessorized with MCR Safety!
: Individually packaged. 100 per box, 10 boxes per case.