Yukon®Visitor Spec, Dielectric, MCR Safety
The Yukon® features a contemporary, dielectric wrap-around style.
- Excellent option for visitor glasses
- Dielectric, no metal parts
- Rugged polycarbonate lens filters 99.9% of harmful UV rays
- Single lens design with integral side shields for greater orbital seal
- Spatula temples
It provides increased angular coverage and improved peripheral vision. Molded-in brow guard and vented sideshields give additional protection for welding. Available in both hard-coated for scratch resistance, or uncoated for extra cost savings.
: Meets or exceeds ANSI Z87+ and CSA Z94.3.
Frequently Bought Together