Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
Complete Certificate of Analysis available for each production lot, along with full formulation information.
It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Caution: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Liquid media products are formulated with cell culture grade WFI water
- Manufactured in a cGMP environment
Complete Certificate of Analysis available for each production lot, along with full formulation information.
It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
Caution: Storage temperature: Solutions 2 - 30 °C; Powder formats 2 - 8 °C
- Catalog No:
- CA45000-426
- CA45000-432
- CA45000-438
- CA45000-440
- CA45000-428
- CA45000-430
- CA45000-434
- CA45000-436
- CA45000-442
- CA45000-444
- Storage temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Refrigerate
- Refrigerate
Product Family Options
DPBS (Dulbecco's Phosphate Buffered Saline)
- DésignationVolumeTempérature de stockage
- DPBS, 10X with calcium and magnesium500 ml+2 °C…+30 °CPack of 6 (500ml)$277.00: 20-030-CV
Specifications
Cat. No.CA45000-426Storage temperatureAmbient - DPBS, 1X with calcium and magnesium1 L+2 °C…+30 °CPack of 6 (1l)$238.00: 21-030-CM
Specifications
Cat. No.CA45000-432Storage temperatureAmbient - DPBS, 1X without calcium and magnesium10 L+2 °C…+30 °CEach (10l)$526.00: 21-031-LX
Specifications
Cat. No.CA45000-438Storage temperatureAmbient - DPBS, 1X without calcium and magnesium20 L+2 °C…+30 °CEach (20l)$745.00: 21-031-LB
Specifications
Cat. No.CA45000-440Storage temperatureAmbient - DPBS, 10X without calcium and magnesium500 ml+2 °C…+30 °CPack of 6 (500ml)$352.00: 20-031-CV
Specifications
Cat. No.CA45000-428Storage temperatureAmbient - DPBS, 1X with calcium and magnesium500 ml+2 °C…+30 °CPack of 6 (500ml)$151.00: 21-030-CV
Specifications
Cat. No.CA45000-430Storage temperatureAmbient - DPBS, 1X without calcium and magnesium500 ml+2 °C…+30 °CPack of 6 (500ml)$138.00: 21-031-CV
Specifications
Cat. No.CA45000-434Storage temperatureAmbient - DPBS, 1X without calcium and magnesium1 L+2 °C…+30 °CPack of 6 (1l)$410.00: 21-031-CM
Specifications
Cat. No.CA45000-436Storage temperatureAmbient
DPBS powder format
- DésignationVolumeTempérature de stockage
- DPBS, Powder without calcium and magnesium10 L+2…+8 °CEach (10l)$48.00: 55-031-PB
Specifications
Cat. No.CA45000-442Storage temperatureRefrigerate - DPBS, Powder without calcium and magnesium50 L+2…+8 °CEach (50l)$66.00: 55-031-PC
Specifications
Cat. No.CA45000-444Storage temperatureRefrigerate