Disposable Heel Grounder, STATICO | Avantor
Disposable Heel Grounder, STATICO
Disposable Heel Grounders
Catalog # 14223-828
Supplier:   Statico
Fournisseur #:  S1100
Disposable Heel Grounder, STATICO
Catalog # 14223-828
Supplier:  Statico
Disposable Heel Grounder, STATICO
Disposable Heel Grounder, STATICO
Disposable Heel Grounders
Price
255,16 $
Log in pour la disponibilité
Pack of 100
Product Details & Documents
The S1100 Series is an easy to use disposable heel grounder that dissipates the static charges effectively, while offering convenience of use
It is designed for use by visitors and mobile personnel at the static sensitive work areas where protection against ESD (Electro Static Discharge) is required. Use of two-foot grounders is required for dependable protection and to meet most ESD control standards.
Certifications: Meets ANSI/ESD S1.1-2013 & ANSI/ESD S20.20-2014.
Ordering information: Heel grounders are packaged 100 per pack.
Spécifications
  • Désignation:
    Disposable Heel Grounders