Analog Surface Resistivity/Resistance Test Kit, STATICO
14223-824
: Statico
- Description:Analog Surface Resistivity/Resistance Test Kit
- Cat. no.:14223-824
- Supplier No.:S2020A
S2020A is an analog pocket size, light weight, auto ranging (10V to 100V) surface resistivity/resistance indicator.
It can be used as a handheld tester, with two-parallel bars on the back performing the test. It can also be used with the two external 5lb probes.
: NIST Certified.
: Kit includes 1 Each Tester, 1 Each Leather-like Carrying Case, 1 Each 9V Battery, 2 Each Connecting Cords (Tester to Probes), 2 Each 5 lb Probes. All are housed in a heavy duty hinged plastic case with foam insterts.