Dominator Pants, Bib style, MCR Safety
Supplier: MCR Safety
Certificates
About this item
The Dominator Series is made tough and roomy for dirty jobs
- 388BF bib style pant has a plain front (no fly opening), full elastic suspenders for comfort and ease of movement, and take-up snaps at the ankles and waist
- Addditional 10" x 10" reinforced crotch area adds extra durablilty
- Material is treated to be limited flammability
It is easily identified by its kelly green color and has additional features to help protect against splash, grit, and grime.
Specifications
- Catalog No:
- 80082-332
- 80082-330
- 80082-336
- 80082-340
- 80082-338
- 80082-344
- 80082-342
- Garment application:
- Industrial, Outdoor
- Industrial, Outdoor
- Industrial, Outdoor
- Industrial, Outdoor
- Industrial, Outdoor
- Industrial, Outdoor
- Industrial, Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- PVC/polyester/PVC
- PVC/polyester/PVC
- PVC/polyester/PVC
- PVC/polyester/PVC
- PVC/polyester/PVC
- PVC/polyester/PVC
- PVC/polyester/PVC
- Thickness:
- 0.42 mm
- 0.42 mm
- 0.42 mm
- 0.42 mm
- 0.42 mm
- 0.42 mm
- 0.42 mm