PVC/Polyester Garment, Detachable Hood, Snap Front, MCR Safety
The Classic Series Material is the most popular in the Industry
- Jacket features a snap front, take-up snaps at the wrists, detachable hood with drawstring, and two front patch pockets
- Bib pants feature a fly front with snap and take-up snaps at the waist and ankles
It combines the strength and waterproof qualities of PVC with the durability of a polyester backing. The 2003 yellow three piece suit features a jacket with detachable hood and bib style pants.
- Catalog No:
- 80082-270
- 80082-266
- 80082-264
- 80082-258
- 80082-256
- 80082-262
- 80082-260
- 80082-272
- 80082-254
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Industrial, Laboratory
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- PVC/polyester
- PVC/polyester
- PVC/polyester
- PVC/polyester
- PVC/polyester
- PVC/polyester
- PVC/polyester
- PVC/polyester
- PVC/polyester
- Thickness:
- 0.35 mm
- 0.35 mm
- 0.35 mm
- 0.35 mm
- 0.35 mm
- 0.35 mm
- 0.35 mm
- 0.35 mm
- 0.35 mm
Frequently Bought Together