Corning® Accutase® Cell Detachment Solution, Corning | Avantor
Corning® Accutase® Cell Detachment Solution, Corning
Accutase Cell Detachment Solution, 100 ml
Catalog # CA10215-416
Supplier:   Corning
Fournisseur #:  25-058-CI
Corning® Accutase® Cell Detachment Solution, Corning
Catalog # CA10215-416
Supplier:  Corning
Corning® Accutase® Cell Detachment Solution, Corning
Corning® Accutase® Cell Detachment Solution, Corning
Accutase Cell Detachment Solution, 100 ml
Price
86,55 $
Log in pour la disponibilité
Each (100ml)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Accutase® is an effective cell detachment solution for the routine detachment of cells from standard tissue culture plasticware as well as enhanced surface and advanced surface coated cultureware
  • Efficient, with gentle detachment of adherent cells within minutes
  • An alternative to Trypsin
  • Able to eliminate neutralization or wash steps
  • Convenient and cost effective
Accutase® is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting. It contains proprietary marine-origin enzymes with proteolytic and collagenolytic activity for the detachment of primary and stem cell lines and tissues. Accutase® provides a reduced contamination risk, because it is free of mammalian or bacteria-derived products.
Spécifications
  • Volume:
    100 ml
  • Désignation:
    Accutase Cell Detachment Solution