Two-Layer Rubber ESD Mat, STATICO
À propos de cet article
The S1400 Series is a two-layer, heat and chemical resistant rubber matting
This product is ideal for anti static workstations where repair work and soldering is performed. The first layer, available in Blue or Gray with Paper Finish, is a static dissipative work surface and the second layer, available only in black with diamond finish, is a conductive one. The 1400 series provides the highest quality rubber matting in the industry.
Certifications: Meets ANSI/ESD S20.20-2014.
Ordering information: Includes one (1) 4.6m (15 ft.) Mat/Wrist Strap Common Point Grounding Cord and two (2) 10mm (3/8 in.) Male Studs.
9 Options Available Below
Product Details & Documents
The S1400 Series is a two-layer, heat and chemical resistant rubber matting
This product is ideal for anti static workstations where repair work and soldering is performed. The first layer, available in Blue or Gray with Paper Finish, is a static dissipative work surface and the second layer, available only in black with diamond finish, is a conductive one. The 1400 series provides the highest quality rubber matting in the industry.
Certifications: Meets ANSI/ESD S20.20-2014.
Ordering information: Includes one (1) 4.6m (15 ft.) Mat/Wrist Strap Common Point Grounding Cord and two (2) 10mm (3/8 in.) Male Studs.
Product Family Options
Informations sur le produit
- ColorIncludesTailleAvailabilityPrice
- BlueStuds Installed61 cm x 91 cm x 2.03 mm (24" x 36" x 0.080")Discontinued-Supplier Number: -
- GrayStuds Installed61 cm x 91 cm x 2.03 mm (24" x 36" x 0.080")Log in pour la disponibilitéEach$414.32Supplier Number: S1460-2436GYWS
- BlueStuds Installed61 cm x 122 cm x 2.03 mm (24" x 48" x 0.080")Log in pour la disponibilitéEach$532.23Supplier Number: S1460-2448BBWS
- GrayStuds Installed61 cm x 122 cm x 2.03 mm (24" x 48" x 0.080")Log in pour la disponibilitéEach$529.34Supplier Number: S1460-2448GYWS
- BlueStuds Uninstalled61 cm x 15.25m x 2.03 mm (2' x 50' x 0.080")Log in pour la disponibilitéEach$4,561.05Supplier Number: S1460-24600BB
- GrayStuds Uninstalled61 cm x 15.25m x 2.03 mm (2' x 50' x 0.080")Log in pour la disponibilitéEach$4,561.05Supplier Number: S1460-24600GY
- BlueStuds Uninstalled76 cm x 15.25m x 2.03 mm (2.5' x 50' x 0.080")Log in pour la disponibilitéEach$5,696.13Supplier Number: S1460-30600BB
- GrayStuds Uninstalled76 cm x 15.25m x 2.03 mm (2.5' x 50' x 0.080")Log in pour la disponibilitéEach$5,696.13Supplier Number: S1460-30600GY
- BlueStuds Uninstalled92 cm x 15.25m x 2.03 mm (3' x 50' x 0.080")Log in pour la disponibilitéEach$6,734.70Supplier Number: S1460-36600BB
- GrayStuds Uninstalled92 cm x 15.25m x 2.03 mm (3' x 50' x 0.080")Log in pour la disponibilitéEach$8,906.06Supplier Number: S1460-36600GY