Kettle Brushes and Covers, Micronova
Supplier: Micronova
Certificates
About this item
The Kettle brush is an oblong-shaped brush with a flare head design and polyester bristles. It is well suited for an all-around, thorough cleaning of small tanks, carboys, and other production equipment.
- Lightweight, durable brush with optional fabric covers
- Made from FDA approved, BPA-free base resins
- FDA approved for indirect food contact
- ACME threaded handle attachment
Attaches to Micronova's STA series, fixed and extendible length handles, to reach the bottom of large process tanks and kettles. The large brush features three handle attachments; one at 90° and two angled on either side fore more versatile cleaning.
Ordering information: Optional covers and handles sold separately.
Frequently Bought Together