Plastic Squeegees, Micronova
These durably constructed squeegees are made with a polypropylene frame and a choice of either solid rubber or double foam blades for use in wash and production areas. Designed with an ACME threaded handle connection for use with most threaded plastic handles.
- Choice of solid rubber or double foam blades
- Integrated blade eliminates seams and crevices, reducing areas for bacteria to grow
- Excellent for use on smooth surfaces and uneven floor surfaces
- Prevents hazardous sparks when cleaning up flammable fluids
- Generous 24" width - ideal for large production areas
- Designed for use with STAH or STAEH handles
- Available in a variety of colors
- Will not rust
The Micronova Plastic Squeegees easily clean smooth or uneven surfaces and minimize contamination areas by reducing dirt and bacteria growth. For use with HACCP compliance programs and color coding.
Frequently Bought Together