SwabTool™ Swab Holder, Micronova
10753-252
Small Business Enterprise
- Description:SwabTool™ adapter
- Material:Stainless steel 304, electropolished, acetal components
- Cat. no.:10753-252
- Supplier No.:QDSWT-01
A custom tool specifically designed to hold swabs for environmental monitoring and sampling. Use with SSU series handles for extension in large tanks and other hard to reach areas.
- Swab is locked in place with a twist knob
- Adapted to swabs with a 6.4 mm (1/4 in) handle
- Optional 'straws' available to hold smaller or longer swabs
- Crafted from electropolished stainless steel with acetal components
- Quick Disconnect fitting connects to SSU series stainless steel handles
- Compatible with strong disinfectants
- Autoclavable
The Micronova SwabTool™ Adapter is compatible with a range of swabs when used with optional 'straws' to hold those which aresmaller or longer. Choose from the range of SSU series handles to match the length required for reach.