Plastic ACME Threaded Handles, Micronova
Supplier: Micronova
Certificates
About this item
Lightweight, plastic coated fiberglass handles with threaded ends for use with the entire range of Micronova adapters, mop heads and tools for simplified and versatile cleaning.
- Lightweight, plastic coated fiberglass handles
- ACME Connect threaded end connection
- Hand grips available in four color options for color coding
- Available fixed length or extendable
- Fixed lengths range from 12 to 60 inches (30 cm to 1.5 m)
- Extendable maximum lengths range from 36 in to 11.5 ft (90 cm to 3.5 m)
- ACME Connect threaded end attaches directly to plastic threaded mops, tools and adapters
- SSA-2 adapter connects threaded handles to Quick Disconnect mops, tools and adapters
- Good compatibility with disinfectants
- Autoclavable
The Micronova Plastic ACME Threaded Handles attach directly to the plastic (QP series) adapters and tools. Use the SSA-2 ACME Thread Adapter to attach to stainless steel Quick Disconnect (QD series) adapters and tools.