Ninja X® Gloves, MCR Safety
Fournisseur: MCR Safety
Certificats
À propos de cet article
Each of the offerings in the Ninja® series include a unique shell and polymer combination incorporating the latest in hand protection technology.
- Available in sizes small through extra large
Our Ninja® Glove Line offers the greatest assortment of dexterity, sense of touch, and protection. Common features to all styles include individual packaging for point of sale display, treated with Actifresh® to kill bacteria and promote freshness.
Spécifications
- Catalog No:
- 89074-246
- 89074-244
- 89074-242
- 89074-240
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Gauge:
- 15 Gauge
- 15 Gauge
- 15 Gauge
- 15 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist