FlexTherm® Gloves, MCR Safety
Fournisseur: MCR Safety
Certificats
À propos de cet article
Flex Therm options give all of the benefits of flex tuff series of gloves, but with the addition of Thermal protection! The 9690 is made of a an acrylic/polyester/cotton string knit shell with a gray latex palm coating.
- Extra thermal protection
- High comfort level
- Allows manual dexterity
- For heavier duty requirements
These gloves can provides a grip on staying warm.
Ordering information: These gloves are polybagged by the pair.
Spécifications
- Catalog No:
- 80081-718
- 80081-716
- 80081-714
- 80081-712
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Gauge:
- 10 Gauge
- 10 Gauge
- 10 Gauge
- 10 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist