UltraTech® Foam Gloves, MCR Safety
Supplier: MCR Safety
Certificates
About this item
The UltraTech® series provides users with tactile sensitivity, dexterity, and abrasion resistance.
- Provides good grip in wet or dry applications
- Excellent comfort, fit and grip
- Greater dexterity
- Nitrile foam coated palm
This series provides a variety of palm coatings over a durable nylon or poly/cotton shell to fit your needs. The 9674 'UltraTech® Foam' features a 15 gauge 100% nylon shell. CE score of 3121.
Ordering information: These gloves are polybagged by the pair.
Specifications
- Catalog No:
- 89087-946
- 89087-948
- 89087-942
- 89087-944
- 89087-950
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Coated Palm
- Gauge:
- 15 Gauge
- 15 Gauge
- 15 Gauge
- 15 Gauge
- 15 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist