String Knit Gloves, MCR Safety
Supplier: MCR Safety
Certificates
About this item
Our multi-purpose string knit gloves come in various blends and weights.
- 7 gauge heavy weight gray cotton/polyester
- Reversible pattern
- Color coded hem to identify sizes
- Made with FDA accepted materials
Economy, regular, and heavy weight gauge offerings are available. These gloves are suitable for general hand protection in light duty applications.
Specifications
- Catalog No:
- 80081-658
- 80081-654
- 89135-734
- 89135-732
- 89135-736
- 80081-662
- 80081-660
- 80081-664
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Finish:
- Dots On 2 Sides
- Dots On 2 Sides
- Dots On 2 Sides
- Gauge:
- 13 Gauge
- 7 Gauge
- 10 Gauge
- 10 Gauge
- 13 Gauge
- 7 Gauge
- 7 Gauge
- 7 Gauge
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist