Durashield™, Nitrile Gloves, Industry Standard Food Grade, Powder-Free, MCR Safety
Nitrile gloves are made of synthetic latex
- Reversible
- Contain no latex proteins
- Offer excellent resistance to punctures and tears
Nitrile gloves are three times more puncture resistant than rubber and can be used to offer superior resistance to many types of chemicals. This glove is an excellent alternative for individuals who experience allergic reactions to latex. The 6002 Durashield nitrile disposable gloves are 4 mil, textured, have a rolled cuff, and are powder-free.
: These gloves are packaged as 100 per dispenser.
- Catalog No:
- 89093-572
- 89093-570
- 89093-568
- 89093-566
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Rolled
- Rolled
- Rolled
- Rolled
- FDA regulated:
- +
- +
- +
- +
Frequently Bought Together