Latex Gloves, Flock Lined, Industrial Grade, MCR Safety
Unsupported gloves are used for a wide variety of applications.
- Scalloped cuff
Flock lining allows for easy on, easy off as well as perspiration absorption. Flock lined latex gloves are made with component materials that comply with FDA regulations for food contact.
: These gloves are polybagged by the pair.
- Catalog No:
- 89087-888
- Ambidextrous/Hand-specific:
- Ambidextrous
- Latex/Latex-free:
- Latex
- Liner material:
- Flock
- Cuff style:
- Straight
- FDA regulated:
- +
Frequently Bought Together