Latex Canner Gloves, Premium Rolled Cuff, MCR Safety
Fournisseur: MCR Safety
Certificats
À propos de cet article
These glove features a premium 18 mil latex canners glove.
- High quality latex ensures high performance on the job
- Don these gloves for extra protection
Amber in color and has a rolled cuff.
Ordering information: These gloves are poly bagged by the pair.
Spécifications
- Catalog No:
- 80081-418
- 80081-420
- 80081-424
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Cuff style:
- Rolled
- Rolled
- Rolled
- FDA regulated:
- +
- +
- +