Portable ESD Field Service Kit, STATICO
Catalog # 14223-740
Supplier: Statico
Specifications
- Description:Portable Field Service Kit
- Cat. no.:14223-740
- Supplier No.:S1920
Specifications
About this item
STATICO's Portable Field Service Kits are designed to protect ESD (Electro Static Discharge) sensitive components and devises at remote locations, where static free workstations are not available
The S1920 is the ideal means for field service operators, safeguarding against ESD damage.
Certifications: Meets ANSI/ESD 20.20-2014.