Double Stack Connector, Statico
Double Stack Connector, Yellow, 10 mm (3/8")
14223-736
: Statico
: S1092DS
Molded male/female double-stack snap with banana jack and caps. Provides a connecting point for:
- Wrist grounding strap via banana jack
- Snaps-on table mat either via female or male 3/8" (10 mm) socket
Either one of the unused snaps can be covered by a plastic cap, to prevent shock due to exposure.
- Désignation:Double Stack Connector, Yellow, 10 mm (3/8")