Adjustable Heel Grounder, STATICO
Adjustable Heel Grounder
Catalog # 14223-706
Supplier: Statico
Fournisseur #: S1150HG
Jump to:
Product Details & Documents
This Adjustable Heel Grounder is a cup design Heel Grounding Strap that provides the end user with an effective means of charge dissipation in the static protected areas
This product is made out of a 2-layer rubber material. Inner layer is beige and static dissipative, and the outer layer is conductive in black color. Adjustment is made easy through an elastic hook and non-elastic loop Blue material. The product has a 22” long conductive tab for skin contact and a sewn-in 1 Meg current limiting safety resistor.
: Meets ANSI/ESD 20.20-2014.
Spécifications
- Désignation:Adjustable Heel Grounder