BEMCOT M-3 Wipes, High-Tech Conversions
BEMCOT Nonwoven, Quarter-Fold Wipes, 25×25 cm (10×10")
Catalog # 10770-150
Small Business Enterprise
Supplier: High-Tech Conversions
Fournisseur #: M-3
BEMCOT M-3 Wipes, High-Tech Conversions
Catalog # 10770-150
Supplier: High-Tech Conversions
Small Business Enterprise
Jump to:
Product Details & Documents
BEMCOT wipes are used in cleaning heat-resistant equipment and diffusion furnaces. They can also be used on sensitive electrical equipment in cleanroom. Made of 100% filament rayon.
- Super absorbent, antistatic
- Nonwoven
- Quarter folded
- Heat resistant
- Double bagged
- Resistant to solvents
Ordering information: Wipes are packaged 100 wipes per bag with 30 bags per case.
Spécifications
- Désignation:BEMCOT Nonwoven, Quarter-Fold Wipes
- Dimensions:25×25 cm (10×10")
- Cat. No.:10770-150
- Basis weight:27.5 g/m²