Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Somso® Thoracic Vertebral Column | Avantor

Somso® Thoracic Vertebral Column

Thoracic Vertebral Column, 18 cm, 18 cm, 37 cm
Catalog # 470102-768
Supplier:   MARCUS SOMMER
Fournisseur #:  QS 63
Somso® Thoracic Vertebral Column
Catalog # 470102-768
Supplier:  MARCUS SOMMER
Somso® Thoracic Vertebral Column
Somso® Thoracic Vertebral Column

Thoracic Vertebral Column, 18 cm, 18 cm, 37 cm

Price
$452.00
Log in pour la disponibilité
Each
Product Details & Documents

Cast from a natural specimen, in SOMSO-Plast®.

  • Shows cord and nerve endings
  • Five year warranty

Mounted and flexible with spinal cord and nerve endings. On a stand and green base.

 : This item is a special order item. Available stock may be limited and additional lead time may be required.

Spécifications
  • Désignation:
    Thoracic Vertebral Column
  • Hauteur:
    37 cm
  • Longueur:
    18 cm
  • Width:
    18 cm