Ceramic Rods, Round, Solid, AL23, Thermo Scientific
À propos de cet article
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
27 Options Available Below
Product Details & Documents
Pure Al2O3 Aluminium oxide, AL23
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Solid aluminum rod.
Product Family Options
Informations sur le produit
- LongueurØAvailabilityPrice
- 400 mm1.5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.AA32925-QAT - 400 mm2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.AA32926-QAT - 400 mm3 mmDiscontinued-Supplier Number: 032927-QATSpécifications:
More
Specifications
Cat. No.AA32927-QAT - 400 mm4 mmDiscontinued-Supplier Number: 032929-QATSpécifications:
More
Specifications
Cat. No.AA32929-QAT - 400 mm5 mmDiscontinued-Supplier Number: 032931-QATSpécifications:
More
Specifications
Cat. No.AA32931-QAT - 400 mm6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.AA32933-QAT - 400 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.AA32935-QAT - Supplier Number: 32920-DMSpécifications:
More
Specifications
Cat. No.CAAA32920-DM - Supplier Number: 32920-DRSpécifications:
More
Specifications
Cat. No.CAAA32920-DR - 250 mm0.5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32920-DT - Supplier Number: 32920-DWSpécifications:
More
Specifications
Cat. No.CAAA32920-DW - 100 mm0.6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32921-DM - 200 mm0.6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32921-DR - 300 mm0.6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32921-DW - Supplier Number: 32922-DMSpécifications:
More
Specifications
Cat. No.CAAA32922-DM - Supplier Number: 32922-DRSpécifications:
More
Specifications
Cat. No.CAAA32922-DR - Supplier Number: 32922-DTSpécifications:
More
Specifications
Cat. No.CAAA32922-DT - Supplier Number: 32922-DWSpécifications:
More
Specifications
Cat. No.CAAA32922-DW - 100 mm1.0 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32923-DM - 200 mm1.0 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32923-DR - 250 mm1.0 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32923-DT - Supplier Number: 32923-DWSpécifications:
More
Specifications
Cat. No.CAAA32923-DW - 100 mm1.2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32924-DM - 200 mm1.2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32924-DR - 250 mm1.2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32924-DT - Supplier Number: 32924-DWSpécifications:
More
Specifications
Cat. No.CAAA32924-DW - Supplier Number: 32925-DMSpécifications:
More
Specifications
Cat. No.CAAA32925-DM - Supplier Number: 32925-DRSpécifications:
More
Specifications
Cat. No.CAAA32925-DR - 250 mm1.5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32925-DT - 300 mm1.5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32925-DW - Supplier Number: 32926-DMSpécifications:
More
Specifications
Cat. No.CAAA32926-DM - 200 mm2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32926-DR - Supplier Number: 32926-DTSpécifications:
More
Specifications
Cat. No.CAAA32926-DT - 300 mm2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32926-DW - 100 mm3 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32927-DM - 200 mm3 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32927-DR - 250 mm3 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32927-DT - 300 mm3 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32927-DW - Supplier Number: 32927-EHSpécifications:
More
Specifications
Cat. No.CAAA32927-EH - 800 mm3 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32927-EM - Supplier Number: 32927-EOSpécifications:
More
Specifications
Cat. No.CAAA32927-EO - Supplier Number: 32929-DMSpécifications:
More
Specifications
Cat. No.CAAA32929-DM - 200 mm4 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32929-DR - Supplier Number: 32929-DTSpécifications:
More
Specifications
Cat. No.CAAA32929-DT - 300 mm4 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32929-DW - 600 mm4 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32929-EH - 800 mm4 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32929-EM - Supplier Number: 32929-EOSpécifications:
More
Specifications
Cat. No.CAAA32929-EO - Supplier Number: 32931-DMSpécifications:
More
Specifications
Cat. No.CAAA32931-DM - Supplier Number: 32931-DRSpécifications:
More
Specifications
Cat. No.CAAA32931-DR - Supplier Number: 32931-DTSpécifications:
More
Specifications
Cat. No.CAAA32931-DT - Supplier Number: 32931-DWSpécifications:
More
Specifications
Cat. No.CAAA32931-DW - Supplier Number: 32931-EHSpécifications:
More
Specifications
Cat. No.CAAA32931-EH - 800 mm5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32931-EM - 1000 mm5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32931-EO - Supplier Number: 32933-DMSpécifications:
More
Specifications
Cat. No.CAAA32933-DM - Supplier Number: 32933-DRSpécifications:
More
Specifications
Cat. No.CAAA32933-DR - 250 mm6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32933-DT - Supplier Number: 32933-DWSpécifications:
More
Specifications
Cat. No.CAAA32933-DW - 600 mm6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32933-EH - Supplier Number: 32933-EMSpécifications:
More
Specifications
Cat. No.CAAA32933-EM - 1000 mm6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32933-EO - 100 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-DM - 200 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-DR - 250 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-DT - 300 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-DW - 600 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-EH - 800 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-EM - 1000 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-EO - 100 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-DM - 200 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-DR - 250 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-DT - 300 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-DW - 600 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-EH - 800 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-EM - 1000 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-EO - 400 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.AA32937-QAT - 400 mm1.5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32925-QAT - 400 mm2 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32926-QAT - 400 mm3 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32927-QAT - 400 mm4 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32929-QAT - 400 mm5 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32931-QAT - 400 mm6 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32933-QAT - 400 mm8 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32935-QAT - 400 mm10 mmDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CAAA32937-QAT