ContactUsLinkComponent
Recharge™ without Glucose and L-Glutamine, Life Technologies
Catalog # CA97060-916
CAS Number:  
undefined
Recharge™ without Glucose and L-Glutamine, Life Technologies
Catalog # CA97060-916
Supplier Number:  670003
CAS Number:  
Specifications
  • Pack type:
    Bottle
  • Size:
    1 kg
  • Cat. no.:
    CA97060-916
  • Supplier No.:
    670003
Specifications
About this item
Recharge™ is a fully chemically defined, animal-free, and protein-free cell culture supplement designed to be used in chemically defined media.