Biox Cryogenic Storage Boxes | Avantor
Biox Cryogenic Storage Boxes
Biox Cryogenic Storage Boxes
Biox Cryogenic Storage Boxes
Biox Cryogenic Storage Boxes
Cryogenic storage boxes are manufactured from polycarbonate for liquid nitrogen and deep freezer use.
  • Clear removable lid
  • Numbered grip
  • Drain holes
  • 1 and 2" options available
Always in a 10×10 configuration. Openings in the bottom make them scan ready when used with 2D barcoded vials.
Ordering information: Storage boxes are packaged 12 per case.
Product Family Options
  • Nb de places
    Dimensions
    Matière
  • 100
    133L×133W×53H mm
    Polycarbonate
    Case of 12
    Case of 12 - $419.40
    $419.40
    Case of 12
       $419.40
     : BX98-0213
  • 100
    133L×133W×36H mm
    Polycarbonate
    Case of 12
    Case of 12 - $409.86
    $409.86
    Case of 12
       $409.86
     : BX98-0113