InLab® OptiOx Dissolved Oxygen Sensors, METTLER TOLEDO®
Fournisseur: Mettler Toledo
Certificats
À propos de cet article
Inlab® OptiOx dissolved oxygen electrode is based on the reliable RDO® technology
- RDO® technology
- For use in harsh environments
- Integrated temperature sensor
Ideal for use in harsh and field based environment such as quality control. Features integrated temperature sensor (NTC 30 KΩ), mini-LTW connector and PC/ABS shaft.
Frequently Bought Together