Shipper, EHS Frozen, Therapak®
EHS frozen shipping system
Catalog # 10029-404
Supplier: Therapak, LLC
Fournisseur #: 56877
Jump to:
Product Details & Documents
EHS shipping systems for frozen specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and box (OD: 11 x 9 x 11”) with a Class 9 Dry Ice label, making it ideal for overnight shipments.
Includes specimen bag, absorbent material, Exempt Human Specimen label, biohazard symbol label, and cushioning material. Add sample tubes and dry ice to ship.
Spécifications
- Désignation:EHS frozen shipping system