Shipper, Multisample EHS Ambient, Therapak®
Catalogue # 10029-400
Fournisseur: Therapak, LLC
Spécifications
- Description:Large transport box, multi-sample shipper
- Cat. no.:10029-400
- Supplier No.:56875
Spécifications
À propos de cet article
Large capacity ambient shipping system accommodates multiple specimen transport bags for consolidation at the collection site and shipment to testing laboratory
System includes a large transport box (ID: 11 x 10.5 x 4"; OD: 11.25 x 10.75 x 4.25") with easy lock flaps on both ends of the box, a 12 x 18" biohazard transport bag, 12 x 12" absorbent sheet. Holds up to ten 6 x 9" specimen transport bags.