ContactUsLinkComponent
Shipper, EHS Ambient, Therapak®
Catalogue # 10029-394
Fournisseur:  Therapak, LLC
undefined
Shipper, EHS Ambient, Therapak®
Catalogue # 10029-394
Fournisseur:  Therapak, LLC
Supplier Number:  56872
Spécifications
  • Description:
    Small transport box, specimen bag and absorbent
  • Cat. no.:
    10029-394
  • Supplier No.:
    56872
Spécifications
À propos de cet article
Ambient shipping system is designed for shipping one standard tube or larger specimen containers (30 mL to 60 mL) of liquid specimen
System features: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, absorbent material, Exempt Human Specimen label, biohazard symbol label and cushioning material where applicable.