TearTech® Shipper, EHS Ambient, Therapak®
Catalog # 10029-392
Supplier: Therapak, LLC
Specifications
- Description:Teartech® EHS ambient specimen shipper
- Cat. no.:10029-392
- Supplier No.:56871
Specifications
About this item
The TearTech® EHS specimen shipper features a bulk-pack case of shipping supplies to transport ambient samples.
The shipper includes 50 easy to assemble TearTech® boxes, 100 zip-style biohazard transport bags (two pocket bag for specimen sample and requisition form), 100 Aqui-Pak™ segmented absorbent pouches (each Aqui-Pak™ holds up to 6 standard blood or urine tubes), 50 biohazard labels, 50 Exempt Human Specimen labels, and cushioning material where applicable.
The system features the TearTech® EHS shipper box that holds up to four specimen bags consisting of patient samples.