Shipper, Category B Combo, Frozen/Ambient, Therapak®
Catalogue # 10029-386
Fournisseur: Therapak, LLC
Spécifications
- Description:Cat B combo, frozen/ambient
- Cat. no.:10029-386
- Supplier No.:56726
Spécifications
À propos de cet article
Features insulated cooler section and ambient section.
- Combination vertical shipper offers the versatility of shipping both ambient and frozen specimens in the same box
- Holds up to six tubes in each section for overnight shipments
The insulated cooler section (ID: 8 x 6 x 8") for frozen specimens includes an Aqui-Pak™ absorbent pouch that will hold up to 6 tubes and a medium 95kPa canister for secondary containment. The ambient section includes an insulated envelope, an Aqui-Pak™ absorbent pouch that will hold up to 6 tubes, a 95 kPa bag for secondary containment and an ambient gel wrap to neutralize temperature spikes during transit. Other materials include orientation labels, list of contents card, biohazard symbol label, frozen specimen label, dry ice label, Biological Substance Category B label and transport box.