VWR® Traceable® Digital pH/ORP Meter | Avantor
VWR® Traceable® Digital pH/ORP Meter
Traceable® pH/ORP meter
Catalog # 10530-802
Supplier:   VWR International
VWR® Traceable® Digital pH/ORP Meter
Catalog # 10530-802
Supplier:  VWR International
VWR® Traceable® Digital pH/ORP Meter
VWR® Traceable® Digital pH/ORP Meter
Traceable® pH/ORP meter
Price
$1,069.81
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
This pH tester is ideal for wastewater, water quality and environmental applications. Compact, water resistant (IP67) instrument fits into the palm of your hand.
  • Compact and portable
  • Water-resistant
  • Extremely accurate
These meters are easy to use: simply dip into solution and read pH or ORP.
Easy-to-operate, always-ready unit quickly recalls exact date and time when minimum and maximum temperatures occur. Audible and visual alarm feature provides alert when temperature rises above or falls below two set points. Alarm is programmable in 1 degree increments.
Portable unit with extreme accuracy (± 0.02 pH)) has a triple display that shows pH or ORP measurement, temperature and time/date. Twenty-five point memory storage with time/date stamp allows for easy recalling of data.
 : To assure accuracy, a Traceable® Certificate is provided from a ISO/IEC 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
 : Supplied with batteries, probe cover, carrying case, detachable probe, Traceable® One-Shot™ pH Buffer Standards (CRM) and a Traceable® Certificate
Spécifications
  • Désignation:
    Traceable® pH/ORP meter
  • Cat. No.:
    10530-802
  • mV range:
    -1800 to 1800mV
  • pH accuracy:
    ± 0.02 pH
  • pH resolution:
    0.01 pH
  • pH range:
    0.00 to 14.00 pH
  • Temperature range:
    0° to 60° C (32° to 140°F)
  • mV accuracy:
    ± 20 mV
  • mV resolution:
    0.1mV < 1000mV, 1mV > 1000mV
  • Temperature resolution:
    0.1° to 100°, 1° above 100°C