ContactUsLinkComponent
Shipper, Category B Refrigerated, Large 95 kPa Canister, Therapak®
Shipper, Category B Refrigerated, Large 95 kPa Canister, Therapak®
Catalog # 10029-296
Supplier:  Therapak, LLC
undefined
Shipper, Category B Refrigerated, Large 95 kPa Canister, Therapak®
Catalog # 10029-296
Supplier:  Therapak, LLC
Supplier Number:  37814
Specifications
  • Description:
    Cat B refrigerated, Large 95kpa canister
  • Cat. no.:
    10029-296
  • Supplier No.:
    37814
Specifications
About this item
Refrigerated Shipping System includes transport box, medium cooler, large 95kPa canister, three Aqui-Pak™ segmented absorbent pouches, 8 oz
refrigerant packs, cushioning material, list of contents card, and required labels for IATA PI650 marking and compliance. Holds up to eighteen tubes for overnight shipments.