ContactUsLinkComponent
Shipper, Category B Frozen, 4 Large Canisters, Therapak®
Catalogue # 10029-274
Fournisseur:  Therapak, LLC
undefined
Shipper, Category B Frozen, 4 Large Canisters, Therapak®
Catalogue # 10029-274
Fournisseur:  Therapak, LLC
Supplier Number:  37809
Spécifications
  • Description:
    Cat B frozen, 4 Large canisters
  • Cat. no.:
    10029-274
  • Supplier No.:
    37809
Spécifications
À propos de cet article
Frozen specimen shipping system includes: transport box (OD: 18x16x14"), large cooler (ID: 16x13x11"), 4 large 95kPa canisters, 12 Aqui-Pak™ segmented absorbent pouches, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance
Holds up to seventy-two tubes for overnight shipments.