Broom and Brush Handles, Perfex
Supplier: Perfex
Certificates
About this item
These handles are lightweighted and ergonomic.
- Polymer handles will not absorb bacteria, liquids or odors
- Constructed from USDA and FDA approved raw materials
- Available with fixed length or adjustable
- Will not rot, corrode or oxidize
- Durable and lightweight (16 oz./64 grams)
- 100% recyclable with unmatched durability
- Available in a variety of sizes and materials
- Easy to clean
Super-smooth finish is pleasant to touch, even in cold conditions. Impervious to water, grease, gasoline, detergents, sanitizers and solvents.
Frequently Bought Together