About this item
Extran® AP 21 The acid special cleaner can be used both as a prewash agent and a rinsing agent with a neutralising effect. When used as a pre-wash agent, it primarily dissolves carbonates and hydroxides from the residues. Protein substances and organic bases, such as amines, are often removed better in an acidic prewash as in an alkaline main wash cycle. As a rinsing agent, i.e. after the alkaline main wash cycle, it is especially suitable for removing remaining traces of alkali on the cleaned material or, in the case of solution carry-over, for neutralisation. This acidic cleaning agent is also well suited to the removal of calcareous deposits in the washing machine.
- Extran® AP 21 is an acidic pre-wash and neutralisation agent with a phosphoric acid base
- Liquid
- Surfactant-free and chlorine-free
- Acidic
- Free from odorants and dyestuffs
Dosing: Added automatically using a dosing device or manually.
- The concentration for application is around0.1–0.3%, i.e. 10–30 mL of Extran® AP 21 are added to around 10 L of water
CAS: 7664-38-2
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA1.07559.9025
- CA1.07559.9010
- CA1.07559.2500
- Formulation:
- Phosphoric acid
- Phosphoric acid
- Phosphoric acid
- pH:
- 2.0 (2 g/l, H₂O, 20 °C)
- 2.0 (2 g/l, H₂O, 20 °C)
- 2.0 (2 g/l, H₂O, 20 °C)
- Solubility:
- (20 °C) Soluble
- (20 °C) Soluble
- (20 °C) Soluble
- Density:
- 1.37 g/cm³ (20 °C)
- 1.37 g/cm³ (20 °C)
- 1.37 g/cm³ (20 °C)