ContactUsLinkComponent
ReliaPrep™ Non-sterile Glass Pre-filter Syringe Filters, Ahlstrom
Catalogue # 10192-196
Fournisseur:  Ahlstrom
undefined
ReliaPrep™ Non-sterile Glass Pre-filter Syringe Filters, Ahlstrom
Catalogue # 10192-196
Fournisseur:  Ahlstrom
Supplier Number:  760529
Spécifications
  • Ø:
    28 mm
  • Pore size:
    0.70 µm
  • Cat. no.:
    10192-196
  • Supplier No.:
    760529
Spécifications
À propos de cet article
Ahlstrom offers syringe filters for convenient filtration of small sample sizes.
This microfiber glass prefilter can be used to remove particles from solution that might block a membrane filter.