BD Eclipse™ Needles and Syringes, BD
À propos de cet article
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
3 Options Available Below
Product Details & Documents
BD Eclipse™ Needles are designed with pivoting needle technology for single-handed activation
For use with BD Luer-Lok™ Syringes.
No hard surface is required with this needle-based, safety-engineered injection device.
Product Family Options
Syringes
Informations sur le produit
- DésignationVolumeJaugeNeedle LengthAvailabilityPrice
- BD Luer-Lok Syringe with Detachable Needle1 ml255/8"Pack of 50$79.50Case of 300$419.22Supplier Number: 305780
- BD Luer-Lok Syringe with Detachable Needle3 ml255/8"Discontinued-Supplier Number: -
- Supplier Number: 305787
- BD Luer-Lok Syringe with Detachable Needle1 ml255/8"Discontinued-Supplier Number: 305780
- BD Luer-Lok Syringe with Detachable Needle3 ml231"Pack of 50$56.60Case of 300$294.96Supplier Number: 305782
- BD Luer-Lok Syringe with Detachable Needle5 ml2211/2"Discontinued-Supplier Number: -
- BD Luer-Lok Syringe with Detachable Needle10 ml2211/2"Discontinued-Supplier Number: -
Needles
Informations sur le produit
- Catalog #LongueurJaugeAvailabilityPrice