À propos de cet article
High-value combination of quality and comfort, as well as reliable cleanroom protection, trust Kimtech™ A5 Nonsterile cleanroom coveralls.
- Breathable SMS fabric
- Clean processed
- High-neck collar for added protection and reduced contamination risk
- Zipper flap to cover the zipper area
- Thumb loops for added protection at the wrist, and to keep sleeves from rolling back
- Bound, tightly sewn seams offer added protection and strengthen barriers
- Bulk packaged for convenience and waste reduction
- Recyclable through the RightCycle™ by Kimberly-Clark Professional®
Made of 100% polypropylene breathable SMS fabric, Kimtech™ A5 Non sterile cleanroom coveralls provide the wearer cloth-like, comfortable fit. While other coveralls for cleanroom environments may look similar, Kimtech™ coveralls offer distinct advantages, including tightly bound seams to strengthen the barrier, a high neck collar to reduce contamination, thumb loops to keep the sleeves from rolling back, and a flap to cover the zipper area.
This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. It is not made with natural rubber latex, silicone, or the BHT preservative. They are designed for single-use (disposable PPE). Typical applications include biotech, medical device manufacturing, electronics, and telecommunications manufacturing.
Spécifications
- Catalog No:
- CA10160-674
- CA10160-672
- CA10160-670
- CA10159-980
- CA10160-678
- CA10160-680
- CA10160-676
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory