KIMTECH PURE® A7 Cleanroom Lab Coat, Kimberly-Clark
Looking for increased protection and superior contamination control? Cover up with the KIMTECH PURE® A7 Cleanroom Lab Coat
- Enhanced chemical and biological protection
- Thumb loops to maintain glove and garment interface
- Low lint to reduce contamination risk
- Advanced Barrier Fabric for better liquid and dry particle protection
- High mandarin collar reduces the risk of contamination
- Elastic wrist and thumb loops for full wrist coverage
- Recyclable through the RightCycle™ by Kimberly-Clark Professional®
- Bulk cleanroom packaging
It all adds up to a safer, greener, more productive cleanroom.
- Catalog No:
- CA10672-492
- CA10672-514
- CA10672-496
- CA10672-510
- CA10672-512
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
Frequently Bought Together