ContactUsLinkComponent
Beaker with Dual Temp Strip, 6.5 oz., Therapak
Beaker with Dual Temp Strip, 6.5 oz., Therapak
Catalog # 10497-208
Supplier:  Therapak, LLC
undefined
Beaker with Dual Temp Strip, 6.5 oz., Therapak
Catalog # 10497-208
Supplier:  Therapak, LLC
Supplier Number:  39416
Specifications
  • Capacity:
    180 ml
  • Description:
    Beaker
  • Cat. no.:
    10497-208
  • Supplier No.:
    39416
Specifications
About this item
This wide-mouth cup is ideal for initial collection of urine samples and subsequent pour off into transport vials.
  • Wide-mouth for easy pouring
  • Sturdy polypropylene construction
  • A variety of configurations to fit your needs
The cup is graduated to 180 ml and its durable, break-resistant design is still flexible enough for squeezing on sides to direct pour off for convenient specimen transfer into transport vial.
Ordering information: This cup is commonly used for collections involving substance abuse testing.