GsBP™ GC columns with high quality column performances in all aspects.
- 100% poly(methylsiloxane) phase
- Equivalent to USP G1, G2, G38
Typical Application: Aldehyde and Ketone, Anabolic Steriods, Anticonvulsants, Aromatics I, Aromatics in Gasoline - ASTM Method 5769, Citronella Java Oil, Common Industrial Solvents, Denatured Fuel Ethanol - ASTM D5501, Diesel Fuel, DNPH Derivative, Drug Screen, EPA Method 551, EPA TO-14 Compounds, Esters I, Ethylene Glycol Mixture, Eucalyptus Oil, Camphor, Menthol, Fatty Acids (Free), Flavor Volatiles, Fragrance Reference Standard I, Freon 12 & Ethylene Oxide, Glycols III, Glycols/Diols, Halogenated Hydrocarbons II, Hydrocarbons Sim-Dis, Inorganic Hydride Gases, Kerosine, Massachusetts APH Mix, Methanol in ethanol, Mineral Spirits, Nitrogen Based Solvent II, Organotin Compounds, Oxygenates in Gasoline ASTM D5599, Ozone Precursors - C2 to C9 Gas Standard, PFBHA Derivative, Polyethylene, Volatile Amines, Pyrethrins, Pyrolysates of Polystyrene, Pyrolysis of High density Polyethylene, Regular Unleaded Gasoline, Solvents IV, Steroids: Sex Hormones, Sulfur in Gasoline, Sulfur in Naphtha, Sulfur Compounds in Natural Gas, Synthetic Essential Oil Mixture, Triethylene Glycol and Impurities.
- Catalog No:
- 10498-832
- 10498-718
- 10499-040
- 10499-042
- 10499-308
- 10498-726
- 10498-084
- 10498-720
- 10498-086
- 10498-722
- 10498-724
- 10498-698
- 10498-690
- 10498-778
- 10498-692
- 10498-694
- 10498-696
- 10498-780
- 10498-782
- 10498-792
- 10499-036
- 10498-794
- 10499-038
- 10499-026
- 10498-784
- 10498-786
- 10499-028
- 10499-292
- 10498-700
- 10498-788
- 10499-294
- 10498-790
- 10499-296
- 10498-676
- 10498-760
- 10498-682
- 10498-770
- 10498-684
- 10498-772
- 10499-014
- 10498-686
- 10498-774
- 10499-016
- 10498-688
- 10498-776
- 10498-762
- 10498-764
- 10498-766
- 10498-768
- Description:
- 10498-832
- 10498-718
- 10499-040
- 10499-042
- 10499-308
- 10498-726
- 10498-084
- 10498-720
- 10498-086
- 10498-722
- 10498-724
- 10498-698
- 10498-690
- 10498-778
- 10498-692
- 10498-694
- 10498-696
- 10498-780
- 10498-782
- 10498-792
- 10499-036
- 10498-794
- 10499-038
- 10499-026
- 10498-784
- 10498-786
- 10499-028
- 10499-292
- 10498-700
- 10498-788
- 10499-294
- 10498-790
- 10499-296
- 10498-676
- 10498-760
- 10498-682
- 10498-770
- 10498-684
- 10498-772
- 10499-014
- 10498-686
- 10498-774
- 10499-016
- 10498-688
- 10498-776
- 10498-762
- 10498-764
- 10498-766
- 10498-768
- Temperature limit:
- -60 to 260/280°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 320/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 320/350°C
- -60 to 300/320°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 300/320°C
- -60 to 380/400°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 360/380°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 300/320°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 325/350°C
- -60 to 325/350°C
- -60 to 260/280°C
- -60 to 300/320°C
- -60 to 260/280°C
- -60 to 260/280°C
- -60 to 280/300°C