GsBP®-VMS Mid-Polar GC Columns, GS-Tek
Fournisseur: General Separation Technologies, Inc.
Certificats
À propos de cet article
Equivalent to: VF-624MS, Rtx-VMS.
- Proprietary phase
- Close to GsBP-1301
- Mid-polar
- USP Code: G43
GC is a common tool for quality control in pharmaceutical production, small molecule research in medical chemistry and drug discovery, metabolism research and other areas. The complexity of these applications demands special columns to conduct separation with a high degree of column inertness and low column bleed for advanced GC, HSGC, HS-GC-MSD and other instrumentation.
Spécifications
- Catalog No:
- 10499-946
- 10499-948
- 10499-950
- Description:
- 10499-946
- 10499-948
- 10499-950
- Temperature limit:
- -20 to 260/280°C
- -20 to 260/280°C
- -20 to 260/280°C