GsBP®-624 Mid-Polar GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
Certificates
About this item
Typical Application: Air, water-drinking, water-underground, water-waste, Packaging, Cosmetics, Healthcare, Drinking, Tobacco, Nutrition, Alcohols, FAME, Halogenated Hydeocarbons, Gasoline, Nitrogen Based Solvents II, Residual Solvents, USP 467, Volatile Amines, Volatile Petroleum Hydrocarbons (VPH), Volatile Sulfurs, Alcohols, DMA, DMSO, Esters, Ethers, Glycols II, EPA 501.3, EPA 503.1, EPA 504, EPA 524.2, EPA 601, EPA 602, EPA 624, EPA 8010, EPA 8015, EPA 8020, EPA 8021, EPA 8240, EPA 8260.
- Proprietary phase
- Close to GsBP-1301
- Mid-polar
- Equivalent to USP G43
Ordering information: These columns are suitable replacements for DB-624, DB-VRX, Rtx-624, Rtx-VMS, DB 502.2, VOCCol.
Specifications
- Catalog No:
- 10499-878
- 10510-868
- 10510-870
- 10510-872
- 10499-920
- 10499-880
- 10499-924
- 10499-882
- 10499-926
- 10499-892
- 10499-896
- 10499-884
- 10499-928
- 10499-886
- 10499-930
- 10499-888
- 10499-932
- 10499-890
- 10499-900
- 10499-902
- 10499-914
- 10499-916
- 10499-906
- 10499-908
- 10499-910
- 10499-912
- Description:
- 10499-878
- 10510-868
- 10510-870
- 10510-872
- 10499-920
- 10499-880
- 10499-924
- 10499-882
- 10499-926
- 10499-892
- 10499-896
- 10499-884
- 10499-928
- 10499-886
- 10499-930
- 10499-888
- 10499-932
- 10499-890
- 10499-900
- 10499-902
- 10499-914
- 10499-916
- 10499-906
- 10499-908
- 10499-910
- 10499-912
- Temperature limit:
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C
- -20 to 260°C