Exonuclease VII
Supplier: New England Biolabs (NEB)
Certificates
About this item
Exonuclease VII, (Exo VII) derived from E.coli, cleaves single-stranded DNA (ssDNA) from both 5´→3´ and 3´→5´ direction. This enzyme is not active on linear or circular dsDNA.
It is useful for removal of single stranded oligonucleotide primers from a completed PCR reaction when different primers are required for subsequent PCR reactions. Digestion of ssDNA by Exonuclease VII is metal-independent.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Enzyme name:Exonuclease VII